Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clca4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207208.3 | NP_997091.3 | 14 | 2480 | ||
GenBank mRNA | AK033591.1 | - | 14 | 2480 | ||
AY560902.2 | - | 2446 | ||||
AY560903.2 | - | 2203 |