Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr153 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178406.2 | NP_848493.1 | 3 | 884 | ||
GenBank mRNA | BC029098.1 | - | 2 | 790 | 3481871 | |
BC037650.1 | - | 2 | 883 | 5151632 |