Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zmym6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285885.1 | NP_001272814.1 | 14 | 3069 | ||
NM_001285887.1 | NP_001272816.1 | 14 | 1986 | |||
NM_001285888.1 | NP_001272817.1 | 15 | 2269 | |||
NM_177462.5 | NP_803413.1 | 15 | 2262 | |||
GenBank mRNA | AK035136.1 | - | 15 | 2264 | ||
AK081836.1 | - | 14 | 1976 | |||
AK170695.1 | - | 15 | 3280 | |||
BC034119.1 | - | 13 | 1914 | |||
BC040744.1 | - | 13 | 1914 | |||
BC094598.1 | - | 14 | 3042 | |||
BC140958.1 | - | 15 | 2137 | 9055570 | ||
BC144807.1 | - | 15 | 2144 |