Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H6pd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291004.1 | NP_001277933.1 | 2 | 521 | ||
NM_173371.4 | NP_775547.2 | 2 | 589 | |||
GenBank mRNA | AK045199.1 | - | 2 | 590 | ||
AK159373.1 | - | 2 | 463 | |||
AK172249.1 | - | 2 | 468 | |||
BC042677.1 | - | 2 | 415 | 4195793 |