Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem64 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181401.3 | NP_852066.2 | 1, 2 | 923 | ||
GenBank mRNA | AK035894.1 | - | 1, 2 | 610 | ||
AK156282.1 | - | 1, 2 | 968 | |||
AK169861.1 | - | 1, 2 | 581 | |||
AK170682.1 | - | 1, 2 | 636 | |||
AY147882.1 | - | 1, 2 | 922 | |||
BC030341.1 | - | 1, 2 | 923 | 3661112 |