Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rusc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037709.2 | NP_001032798.1 | 5 | 3267 | ||
NM_199057.3 | NP_951012.2 | 5 | 3228 | |||
GenBank mRNA | AK053542.1 | - | 2 | 1529 | ||
AK122263.1 | - | 5 | 3147 | |||
AK132170.1 | - | 5 | 3157 | |||
AK166260.1 | - | 5 | 3168 | |||
BC024790.1 | - | 3 | 458 | |||
BC141186.1 | - | 4 | 2998 | 9055798 |