Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rusc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037709.2 | NP_001032798.1 | 4, 5 | 3138 | ||
NM_199057.3 | NP_951012.2 | 4, 5 | 3099 | |||
GenBank mRNA | AK122263.1 | - | 4, 5 | 3018 | ||
AK132170.1 | - | 4, 5 | 3028 | |||
AK166260.1 | - | 4, 5 | 3039 | |||
BC024790.1 | - | 2, 3 | 329 | |||
BC141186.1 | - | 3, 4 | 2869 | 9055798 |