Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stx12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133887.4 | NP_598648.1 | 8 | 779 | ||
GenBank mRNA | AB019211.1 | - | 8 | 722 | ||
AK076390.1 | - | 762 | ||||
AK077678.1 | - | 8 | 780 | |||
AK078948.1 | - | 8 | 739 | |||
AK146544.1 | - | 8 | 784 | |||
AK146735.1 | - | 8 | 771 | |||
AK167087.1 | - | 8 | 693 | |||
AK167804.1 | - | 8 | 743 | |||
AK168753.1 | - | 8 | 738 | |||
AK170203.1 | - | 8 | 816 | |||
BC010669.1 | - | 8 | 766 | 2645010 |