Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Osbpl9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134791.2 | NP_001128263.1 | 14 | 1354 | ||
NM_133885.4 | NP_598646.4 | 17 | 1587 | |||
NM_173350.3 | NP_775485.2 | 15 | 1393 | |||
GenBank mRNA | AK035658.1 | - | 15 | 1342 | ||
AK075981.1 | - | 4 | 372 | |||
AK131984.1 | - | 17 | 1558 | |||
AK172010.1 | - | 15 | 1380 | |||
AK210305.1 | - | 1 | 62 | |||
AK212613.1 | - | 1 | 62 | |||
BC021507.1 | - | 13 | 1215 | 5347121 | ||
BC023759.1 | - | 17 | 1542 | |||
BC026927.1 | - | 15 | 1276 | 5134780 | ||
BC089163.1 | - | 14 | 1295 | 5367603 |