Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp1r8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290725.1 | NP_001277654.1 | 2, 3 | 166 | ||
NM_146154.3 | NP_666266.1 | 2, 3 | 166 | |||
GenBank mRNA | AX775983.1 | - | 2, 3 | 131 | ||
BC025479.1 | - | 2, 3 | 128 | 5354224 | ||
BY257668.1 | - | 2, 3 | 159 | |||
BY293277.1 | - | 2, 3 | 167 | |||
CK625884.1 | - | 2, 3 | 98 |