Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACVR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105.4 | NP_001096.1 | 5 | 868 | ||
NM_001111067.2 | NP_001104537.1 | 5 | 687 | |||
XM_005246939.3 | XP_005246996.1 | 5 | 739 | |||
XM_005246940.3 | XP_005246997.1 | 5 | 707 | |||
XM_006712825.3 | XP_006712888.1 | 5 | 820 | |||
XM_011512106.2 | XP_011510408.1 | 6 | 787 | |||
XM_011512107.2 | XP_011510409.1 | 5 | 759 | |||
XM_011512108.2 | XP_011510410.1 | 5 | 2166 | |||
GenBank mRNA | AK297765.1 | - | 5 | 655 | ||
AK313630.1 | - | 5 | 663 | |||
BC033867.1 | - | 5 | 868 | 5209711 | ||
BC036748.1 | - | 5 | 868 | |||
KU177885.1 | - | 3 | 438 | |||
KU177886.1 | - | 3 | 438 | |||
KU177887.1 | - | 3 | 438 | |||
L02911.1 | - | 6 | 778 | |||
Z22534.1 | - | 4 | 541 |