Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rell1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145923.4 | NP_666035.2 | 6 | 875 | ||
GenBank mRNA | AK133845.1 | - | 7 | 895 | ||
AK140108.1 | - | 6 | 875 | |||
AK159944.1 | - | 6 | 801 | |||
AK161061.1 | - | 6 | 801 | |||
AK209920.1 | - | 3 | 293 | |||
BC023848.1 | - | 2 | 171 | |||
BC024679.1 | - | 3 | 324 | |||
BC031198.1 | - | 6 | 844 | 5059693 | ||
BC066137.1 | - | 6 | 854 | 30012041 | ||
BC066160.1 | - | 6 | 854 | 30012671 |