Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oas1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133893.3 | NP_598654.1 | 5 | 1059 | ||
GenBank mRNA | AB067532.1 | - | 5 | 1013 | ||
AK054465.1 | - | 5 | 1027 | |||
AK135783.1 | - | 4 | 573 | |||
AK136001.1 | - | 5 | 1027 | |||
AY055829.1 | - | 5 | 1021 | |||
BC052835.1 | - | 5 | 1023 | 30048758 |