Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014796.1 | NP_001014796.1 | 11, 12 | 1630 | ||
NM_006182.2 | NP_006173.2 | 10, 11 | 1550 | |||
XM_006711344.3 | XP_006711407.1 | 10, 11 | 1591 | |||
XM_011509586.2 | XP_011507888.1 | 10, 11 | 1523 | |||
XM_011509587.2 | XP_011507889.1 | 11, 12 | 1584 | |||
XM_011509588.2 | XP_011507890.1 | 10, 11 | 1587 | |||
GenBank mRNA | AK225063.1 | - | 10, 11 | 1469 | ||
AK314388.1 | - | 10, 11 | 1499 | |||
BC052998.2 | - | 11, 12 | 1601 | 6162716 | ||
BX640899.1 | - | 11, 12 | 1587 | |||
X74764.1 | - | 10, 11 | 1518 |