Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ugt2b38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133894.2 | NP_598655.1 | 1 | 169 | ||
GenBank mRNA | AK034801.1 | - | 1 | 170 | ||
BC015272.1 | - | 1 | 166 | 4241480 | ||
BC057169.1 | - | 1 | 155 | 4975249 | ||
BC069923.1 | - | 1 | 157 | 4974994 |
Gene Symbol: Ugt2b5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009467.3 | NP_033493.3 | 1 | 171 | ||
GenBank mRNA | AK136480.1 | - | 1 | 172 | ||
BC028262.1 | - | 1 | 162 | 5102040 | ||
X06358.1 | - | 1 | 149 |