Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ugt2b37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053215.3 | NP_444445.2 | 1 | 646 | ||
GenBank mRNA | AK002736.1 | - | 1 | 663 | ||
BC019487.1 | - | 1 | 646 | 4237718 |
Gene Symbol: Ugt2b38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133894.2 | NP_598655.1 | 1 | 660 | ||
GenBank mRNA | AK034801.1 | - | 1 | 661 | ||
BC015272.1 | - | 1 | 657 | 4241480 | ||
BC057169.1 | - | 1 | 646 | 4975249 | ||
BC069923.1 | - | 1 | 648 | 4974994 |