Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc8c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133897.2 | NP_598658.1 | 3, 4 | 367 | ||
GenBank mRNA | AB081508.1 | - | 3, 4 | 367 | ||
AK089024.1 | - | 3, 4 | 435 | |||
AK142337.1 | - | 3, 4 | 397 | |||
AK169492.1 | - | 3, 4 | 430 | |||
AK171313.1 | - | 3, 4 | 449 | |||
BC026572.1 | - | 3, 4 | 377 | 4988595 |