Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Noc4l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153570.2 | NP_705798.2 | 10 | 983 | ||
GenBank mRNA | AK052601.1 | - | 10 | 983 | ||
AK090024.1 | - | 10 | 995 | |||
AK146006.1 | - | 10 | 979 | |||
AK172310.1 | - | 10 | 962 | |||
BC024616.1 | - | 10 | 973 | 4218310 |