Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: N4bp2l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133898.4 | NP_598659.3 | 4, 5 | 548 | ||
GenBank mRNA | AK046116.1 | - | 4, 5 | 525 | ||
AK131647.1 | - | 4, 5 | 540 | |||
AK138666.1 | - | 4, 5 | 524 | |||
BB629746.1 | - | 4, 5 | 548 | |||
BC019754.1 | - | 3, 4 | 510 | 4007588 |