Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Papd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133905.2 | NP_598666.1 | 2 | 312 | ||
GenBank mRNA | AK146086.1 | - | 2 | 296 | ||
AM419011.1 | - | 1 | 85 | |||
BC016629.1 | - | 2 | 312 | 3595512 |