Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Emilin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133918.2 | NP_598679.1 | 2, 3 | 383 | ||
GenBank mRNA | AK029337.1 | - | 2, 3 | 354 | ||
AK052368.1 | - | 2, 3 | 464 | |||
AK088995.1 | - | 2, 3 | 447 | |||
AK155486.1 | - | 2, 3 | 685 | |||
BC005481.1 | - | 2, 3 | 383 | 3485057 |