Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NTSR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002531.2 | NP_002522.2 | 4 | 1794 | ||
GenBank mRNA | X70070.1 | - | 4 | 1795 |
Set Membership: |
G-protein coupled receptor |