Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp282 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146175.3 | NP_666287.3 | 3 | 796 | ||
GenBank mRNA | AK030516.1 | - | 3 | 704 | ||
AK081854.1 | - | 3 | 695 | |||
AK088531.1 | - | 3 | 795 | |||
AK148038.1 | - | 3 | 782 | |||
BC025581.1 | - | 3 | 698 | 5325029 |