Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133924.3 | NP_598685.2 | 1, 2 | 300 | ||
GenBank mRNA | AK017516.1 | - | 1, 2 | 305 | ||
AK141663.1 | - | 1, 2 | 301 | |||
AK168473.1 | - | 1, 2 | 287 | |||
BC117542.1 | - | 1, 2 | 509 | 40105600 |