Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itfg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133927.1 | NP_598688.1 | 2 | 230 | ||
GenBank mRNA | AK134630.1 | - | 2 | 251 | ||
AK142721.1 | - | 2 | 250 | |||
AK147728.1 | - | 2 | 232 | |||
AK159637.1 | - | 2 | 239 | |||
BC014833.1 | - | 2 | 230 | 4507661 | ||
BC025492.1 | - | 2 | 250 | 5309950 |