Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B630005N14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175312.4 | NP_780521.2 | 5 | 946 | ||
GenBank mRNA | AK046745.1 | - | 5 | 947 | ||
AK170526.1 | - | 4 | 2408 | |||
BC023771.1 | - | 1 | 48 | |||
BC145708.1 | - | 5 | 902 | 40131011 | ||
BC145710.1 | - | 5 | 902 | 40131013 |