Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tada3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133932.2 | NP_598693.1 | 3 | 624 | ||
GenBank mRNA | AF383154.1 | - | 3 | 443 | ||
AK078906.1 | - | 3 | 944 | |||
BC023879.1 | - | 4 | 687 | |||
BC026630.1 | - | 3 | 624 | 4983657 | ||
BC032195.1 | - | 4 | 545 | 5323331 |