Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUMA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286561.1 | NP_001273490.1 | 16 | 1986 | ||
NM_006185.3 | NP_006176.2 | 15 | 1888 | |||
XM_006718564.1 | XP_006718627.1 | 14 | 1703 | |||
XM_011545054.1 | XP_011543356.1 | 14 | 1703 | |||
XM_011545055.1 | XP_011543357.1 | 15 | 1881 | |||
XM_011545056.2 | XP_011543358.1 | 16 | 1875 | |||
XM_011545057.2 | XP_011543359.1 | 16 | 1865 | |||
XM_011545058.1 | XP_011543360.1 | 14 | 1902 | |||
XM_011545059.2 | XP_011543361.1 | 16 | 1867 | |||
XM_011545060.2 | XP_011543362.1 | 16 | 1861 | |||
XM_011545061.2 | XP_011543363.1 | 14 | 1721 | |||
XM_011545062.2 | XP_011543364.1 | 14 | 1734 | |||
XM_011545063.2 | XP_011543365.1 | 14 | 1701 | |||
XM_011545064.1 | XP_011543366.1 | 14 | 1688 | |||
XM_011545065.1 | XP_011543367.1 | 14 | 1703 | |||
XM_011545066.1 | XP_011543368.1 | 14 | 1703 | |||
XM_017017830.1 | XP_016873319.1 | 15 | 1743 | |||
XM_017017831.1 | XP_016873320.1 | 14 | 1739 | |||
GenBank mRNA | AB210007.1 | - | 15 | 1689 | ||
AK290509.1 | - | 14 | 1703 | |||
AK307577.1 | - | 15 | 1706 | |||
BC027493.1 | - | 15 | 1689 | |||
BC103765.1 | - | 15 | 1807 | |||
Z11583.1 | - | 15 | 1706 | |||
Z11584.1 | - | 16 | 1808 |