Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccp110 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182995.1 | NP_892040.1 | 5 | 1450 | ||
GenBank mRNA | AK141641.1 | - | 5 | 1445 | ||
AK172946.1 | - | 1 | 664 | |||
BC053103.1 | - | 5 | 1450 | 6406475 |