Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nlrp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133946.2 | NP_598707.2 | 4 | 1588 | ||
GenBank mRNA | AK039531.1 | - | 1 | 872 | ||
AY355343.1 | - | 1 | 526 | |||
BC013519.1 | - | 1 | 495 | |||
BC031139.1 | - | 1 | 580 | 4972485 | ||
FJ222825.1 | - | 4 | 1534 | |||
JF810536.1 | - | 3 | 1973 |