Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ano1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242349.1 | NP_001229278.1 | 18 | 2098 | ||
NM_178642.5 | NP_848757.4 | 19 | 2107 | |||
GenBank mRNA | AK029329.1 | - | 5 | 474 | ||
AK032793.1 | - | 14 | 1119 | |||
AK052589.1 | - | 19 | 2059 | |||
BC004697.1 | - | 11 | 973 | |||
BC006062.1 | - | 2 | 57 | |||
BC062959.1 | - | 18 | 1984 | 30547439 |