Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C230052I12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178643.5 | NP_848758.1 | 2 | 621 | ||
GenBank mRNA | AK031566.1 | - | 2 | 662 | ||
AK040248.1 | - | 2 | 647 | |||
AK082457.1 | - | 2 | 622 | |||
AK153918.1 | - | 2 | 568 | |||
AK166481.1 | - | 2 | 668 | |||
BC096687.1 | - | 2 | 473 | 1433264 |