Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ints4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027256.2 | NP_081532.1 | 23 | 2908 | ||
GenBank mRNA | AK011676.1 | - | 9 | 905 | ||
BC013710.1 | - | 23 | 2854 | 3156446 | ||
BC013813.1 | - | 11 | 1162 | |||
BC021838.1 | - | 16 | 1911 | |||
BC027066.1 | - | 2 | 2763 | |||
BC031145.1 | - | 17 | 1987 |