Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sf3b3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133953.2 | NP_598714.1 | 10, 11 | 1525 | ||
GenBank mRNA | AK040625.1 | - | 10, 11 | 1499 | ||
AK046845.1 | - | 10, 11 | 1510 | |||
AK088268.1 | - | 10, 11 | 1498 | |||
AK128988.1 | - | 10, 11 | 1515 | |||
AK129035.1 | - | 10, 11 | 1493 | |||
AK147914.1 | - | 10, 11 | 1516 | |||
AK164199.1 | - | 10, 11 | 1516 | |||
AK193615.1 | - | 1, 2 | 36 | |||
AK196066.1 | - | 1, 2 | 36 | |||
AK196304.1 | - | 1, 2 | 36 | |||
BC011412.1 | - | 10, 11 | 1506 | 3600115 | ||
BC042580.1 | - | 10, 11 | 1452 | 5367372 | ||
BC139015.1 | - | 9, 10 | 1389 | 8862037 | ||
BC139016.1 | - | 9, 10 | 1389 | 8862038 |