Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ces2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190330.1 | NP_001177259.1 | 3 | 379 | ||
NM_133960.5 | NP_598721.1 | 3 | 379 | |||
GenBank mRNA | AK078953.1 | - | 3 | 375 | ||
AK165784.1 | - | 3 | 369 | |||
BC024082.1 | - | 3 | 345 | 5100146 | ||
BC024491.1 | - | 3 | 354 | 4981579 | ||
BC024517.1 | - | 3 | 352 | 4984109 | ||
BC025537.1 | - | 3 | 365 | 5309880 | ||
BC025812.1 | - | 3 | 352 | 5096915 | ||
BC026643.1 | - | 3 | 352 | 4986124 | ||
BC031295.1 | - | 3 | 345 | 5100144 |