Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nlrc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033207.3 | NP_001028379.2 | 49 | 5742 | ||
GenBank mRNA | AK155962.1 | - | 2 | 526 | ||
BC150954.1 | - | 2 | 520 |