Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dohh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133964.2 | NP_598725.2 | 2 | 326 | ||
GenBank mRNA | AK004052.1 | - | 2 | 292 | ||
AK154104.1 | - | 2 | 327 | |||
BC002295.1 | - | 2 | 243 | 3590525 |