Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam192a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028221.4 | NP_082497.2 | 5 | 677 | ||
GenBank mRNA | AK010053.1 | - | 5 | 642 | ||
AK033988.1 | - | 5 | 608 | |||
AK050704.1 | - | 5 | 612 | |||
AK151603.1 | - | 5 | 631 | |||
BC016088.1 | - | 4 | 417 | 4503452 | ||
BC023158.1 | - | 5 | 516 | 4946201 | ||
BC025472.1 | - | 5 | 653 | 5353148 | ||
BC027060.1 | - | 5 | 639 | 5358022 | ||
BC027818.1 | - | 4 | 445 | 5320967 | ||
BC040818.1 | - | 630 | 4194531 |