Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taf5l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133966.2 | NP_598727.1 | 2 | 191 | ||
GenBank mRNA | AK033477.1 | - | 2 | 219 | ||
AK089463.1 | - | 2 | 271 | |||
AK154832.1 | - | 2 | 271 | |||
BC013550.1 | - | 2 | 191 | 4222283 |