Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133967.3 | NP_598728.1 | 3 | 783 | ||
GenBank mRNA | AK041905.1 | - | 3 | 652 | ||
AK089681.1 | - | 628 | ||||
AK090333.1 | - | 3 | 628 | |||
AK157968.1 | - | 3 | 636 | |||
AK170639.1 | - | 3 | 636 | |||
AK170923.1 | - | 3 | 606 | |||
AK172462.1 | - | 3 | 617 | |||
AK179015.1 | - | 259 | ||||
AK201914.1 | - | 1 | 141 | |||
AK218562.1 | - | 1 | 141 | |||
AY166673.1 | - | 1 | 301 | |||
BC013467.1 | - | 2 | 334 | 4218025 | ||
BC075666.1 | - | 3 | 783 | 30605163 |