Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agpat6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018743.4 | NP_061213.2 | 6 | 1615 | ||
GenBank mRNA | AF015811.1 | - | 3 | 291 | ||
AF406611.1 | - | 5 | 1126 | |||
AK045235.1 | - | 6 | 1509 | |||
AK049675.1 | - | 6 | 1508 | |||
AK077561.1 | - | 6 | 1499 | |||
AK083589.1 | - | 6 | 1509 | |||
AK161270.1 | - | 6 | 1478 | |||
AK162033.1 | - | 3 | 420 | |||
AK167876.1 | - | 6 | 1524 | |||
AK169257.1 | - | 6 | 1509 | |||
AY489184.1 | - | 5 | 684 | |||
BC031767.1 | - | 5 | 969 | 4457846 | ||
BC057860.1 | - | 3 | 289 |