Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agpat6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018743.4 | NP_061213.2 | 4 | 1421 | ||
GenBank mRNA | AF015811.1 | - | 1 | 97 | ||
AF406611.1 | - | 3 | 932 | |||
AK045235.1 | - | 4 | 1315 | |||
AK049675.1 | - | 4 | 1315 | |||
AK077561.1 | - | 4 | 1305 | |||
AK083589.1 | - | 4 | 1315 | |||
AK161270.1 | - | 4 | 1284 | |||
AK162033.1 | - | 1 | 226 | |||
AK167876.1 | - | 4 | 1330 | |||
AK169257.1 | - | 4 | 1315 | |||
AK178773.1 | - | 3 | 410 | |||
AY489184.1 | - | 3 | 490 | |||
BC031767.1 | - | 3 | 775 | 4457846 | ||
BC057860.1 | - | 1 | 95 |