Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpne7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_170684.2 | NP_733785.1 | 15 | 1714 | ||
GenBank mRNA | AI550750.1 | - | 2 | 127 | ||
AK049823.1 | - | 12 | 2856 | |||
AK162900.1 | - | 14 | 2333 | |||
BC119328.1 | - | 15 | 1613 | 8734077 | ||
BC137595.1 | - | 15 | 1613 | 8860614 |