Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpne7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_170684.2 | NP_733785.1 | 2 | 349 | ||
GenBank mRNA | AK049823.1 | - | 2 | 278 | ||
AK162900.1 | - | 2 | 350 | |||
AU079878.1 | - | 2 | 268 | |||
BC119328.1 | - | 2 | 248 | 8734077 | ||
BC137595.1 | - | 2 | 248 | 8860614 |