Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcun1d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024504.2 | NP_001019675.2 | 2, 3 | 551 | ||
NM_001042649.1 | NP_001036114.1 | 2, 3 | 551 | |||
NM_001042650.1 | NP_001036115.1 | 2, 3 | 551 | |||
NM_001042651.1 | NP_001036116.1 | 2, 3 | 551 | |||
GenBank mRNA | AK034394.1 | - | 2, 3 | 259 | ||
AK154550.1 | - | 2, 3 | 557 | |||
AK155726.1 | - | 2, 3 | 267 | |||
AK161694.1 | - | 2, 3 | 256 | |||
BC030839.1 | - | 2, 3 | 259 | |||
BC052676.1 | - | 2, 3 | 554 | |||
BU053156.1 | - | 2, 3 | 459 | |||
CD353600.1 | - | 1, 2 | 52 |