Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcun1d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024504.2 | NP_001019675.2 | 4 | 810 | ||
NM_001042649.1 | NP_001036114.1 | 4 | 810 | |||
NM_001042650.1 | NP_001036115.1 | 4 | 810 | |||
NM_001042651.1 | NP_001036116.1 | 4 | 810 | |||
GenBank mRNA | AK034394.1 | - | 4 | 518 | ||
AK040130.1 | - | 2 | 422 | |||
AK154550.1 | - | 4 | 816 | |||
AK155726.1 | - | 4 | 526 | |||
AK161694.1 | - | 4 | 515 | |||
BC030839.1 | - | 4 | 518 | |||
BC052676.1 | - | 4 | 813 | |||
CA548310.1 | - | 2 | 153 | |||
CD353600.1 | - | 3 | 311 |