Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281974.1 | NP_001268903.1 | 7 | 1249 | ||
NM_001281975.1 | NP_001268904.1 | 7 | 1299 | |||
NM_133971.3 | NP_598732.2 | 6 | 1137 | |||
GenBank mRNA | AK081754.1 | - | 7 | 1297 | ||
AK082875.1 | - | 6 | 1134 | |||
AK084456.1 | - | 7 | 1247 | |||
AK088371.1 | - | 6 | 1087 | |||
AK198185.1 | - | 1 | 14 | |||
AK207506.1 | - | 1 | 14 | |||
BC002198.1 | - | 6 | 1069 | 3489133 |