Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myzap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033208.4 | NP_001028380.1 | 12 | 1358 | ||
GenBank mRNA | AK132167.1 | - | 11 | 4574 | ||
AK146889.1 | - | 12 | 1359 | |||
BC139077.1 | - | 12 | 1358 | 8862099 | ||
BC139078.1 | - | 12 | 1358 | 8862100 | ||
BU073446.1 | - | 4 | 383 |