Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007713.4 | NP_031739.3 | 2 | 895 | ||
GenBank mRNA | AF033565.1 | - | 2 | 171 | ||
AK090215.1 | - | 2 | 136 | |||
AK133936.1 | - | 2 | 141 | |||
AK136481.1 | - | 2 | 158 | |||
AK140015.1 | - | 2 | 138 | |||
AK145880.1 | - | 2 | 150 | |||
AK146942.1 | - | 2 | 898 | |||
AK154003.1 | - | 2 | 144 | |||
AK167300.1 | - | 2 | 897 | |||
BC085084.1 | - | 2 | 138 | 6309020 |