Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007713.4 | NP_031739.3 | 3 | 987 | ||
GenBank mRNA | AF033565.1 | - | 3 | 263 | ||
AK090215.1 | - | 3 | 228 | |||
AK133936.1 | - | 3 | 233 | |||
AK136481.1 | - | 3 | 250 | |||
AK140015.1 | - | 3 | 230 | |||
AK145880.1 | - | 3 | 242 | |||
AK146942.1 | - | 3 | 990 | |||
AK154003.1 | - | 3 | 236 | |||
AK167300.1 | - | 3 | 989 | |||
BC085084.1 | - | 3 | 230 | 6309020 |